Learn More
Medchemexpress LLC Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin | 55216-11-0 | MFCD00010728 | >=98.0% | 1429.54 | C63H112O35 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW145501250MG
Encompass_Preferred
Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin is a fully methylated β-cyclodextrin derivative used as a biochemical reagent for forming host-guest inclusion complexes, improving aqueous solubility of hydrophobic molecules, and acting as a chiral selector in separation methods. It is applied in formulation research, chiral separations, and physicochemical studies of molecular interactions.
  • Fully methylated derivative that promotes strong host-guest interactions.
  • Improves solubility and stability of hydrophobic compounds in water.
  • Acts as a chiral selector for chromatographic and electrophoretic separations.
  • Applicable to formulation and inclusion-complex studies.
  • Provided in laboratory-scale quantities for research use.
Catalog No. 50-002-83816