Learn More
Medchemexpress LLC Heptaminol hydrochloride | 543-15-7 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1231
Encompass_Preferred
Heptaminol hydrochloride (RP-2831) is a vasoconstrictor studied for its use in hypotension, especially orthostatic hypotension. It also acts as a skin cancer proliferation inhibitor by hindering immune inflammation induced by the tumor-promoting factor 12-O-tetradecanoylphorbol-13-acetate (TPA) in an NO-dependent manner. Additionally, it functions as a sympathomimetic amine, exhibiting indirect sympathetic effects, and is an antagonist of catecholamine release and uptake, which can elevate intracellular free calcium levels.
  • Vasoconstrictor for hypotension studies.
  • Inhibits skin cancer proliferation.
  • Acts as a sympathomimetic amine.
  • Antagonist of catecholamine release and uptake.
  • Increases intracellular free calcium levels.
  • Demonstrates anti-proliferative and anti-inflammatory effects.
Catalog No. 50-004-14311