Learn More
Abcam Heptelidic acid (Koningic acid), GAPDH inhibitor, 1MG
Supplier:  Abcam AB1442691MG
Encompass_Preferred
MW 280.32 Da, Purity >95%. Potent, selective, irreversible GAPDH inhibitor. Selective, competitive DNA polymerase inhibitor. Antibiotic agent. Potently inhibits ATP generation in the glycolytic pathway. Inhibits DNA polymerases β, λ and terminal deoxynucleotidyl transferase (TdT) in family X. Inhibits caspase- 3 induction (IC₅₀ = 40 μM). Shows antiapoptotic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3284