Learn More
Cayman Chemical HeptnoIc AcId methyl ester 1g
SDP
Supplier:  Cayman Chemical 336731 G
Encompass_Preferred
An esterified form of heptanoic acid; a target odor compound in Cascade and Willamette hops
Catalog No. 50-294-8197