Learn More
Sino Biological Peptide Substrates: HER2 Substrate
Supplier:  Sino Biological E2758
Encompass_Preferred
The HER2 Substrate synthetic peptide [AAEEIYAARRG] is routinely evaluated as a substrate for HER2.
Catalog No. 50-256-9231