Learn More
Genscript Corporation Her3 HisCynomolgus/Rhesus ma
Supplier:  Genscript Corporation Z03909500
Encompass
Her3 also called ErbB3 is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors Her3 is expressed in keratinocytes melanocytes skeletal muscle cells embryonic myoblasts and Schwann cells Monomeric Her3 serves as a low affinity receptor for the heregulins (HRG)
Catalog No. 50-000-57452