Learn More
Medchemexpress LLC Herbacetin 3,8-O-diglucoside | 99224-12-1 | 626.52 | 5 MG

Supplier: Medchemexpress LLC HYN100425MG
Herbacetin 3,8-O-diglucoside is a flavonol compound found in flax seeds, intended for research use only. This high-purity compound demonstrates antiviral activity against influenza A viruses and is soluble in DMSO, making it suitable for various laboratory applications.
- High purity (99.36%).
- Molecular formula: C27H30O17.
- Appearance: Solid, light yellow to yellow.
- Antiviral activity against influenza A virus H9N2 (EC50: 100 μM).
- Antiviral activity against influenza A virus H1N1 (EC50: 99.1 μM).
- Soluble in DMSO (≥ 125 mg/mL).
- Storage conditions for optimal stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.