Learn More
Medchemexpress LLC HY-N0240 5mg Medchemexpress, Herbacetin CAS:527-95-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02405MG
Encompass_Preferred
Medchemexpress, HY-N0240 5mg Herbacetin CAS:527-95-7 Herbacetin is a natural flavonoid from flaxseed, exerts various pharmacological activities, including antioxidant, anti-inflammatory and anticancer effects. Herbacetin is an Ornithine decarboxylase (ODC) allosteric inhibitor, directly binds to Asp44, Asp243, and Glu384 on ODC. Ornithine decarboxylase (ODC) is a rate-limiting enzyme in the first step of polyamine biosynthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0238