Learn More
Selleck Chemical LLC Herbacetin-E0208-1MG
SDP
Supplier:  Selleck Chemical LLC E02081MG
Encompass_Preferred
Herbacetin is a natural flavonoid compound found in flaxseed with strong antioxidant antitumor and anti-inflammatory activity
Catalog No. 50-313-8516