Learn More
Agilent Technologies HERCULASE ENHANCED DNA POLY
Supplier:  Agilent Technologies 600260
Encompass
Herculase Enhanced DNA Polymerase 100 U Challenging cloning applications Long and/or GC -rich targets Amplifies a wide range of complex targets up to 50 kb Higher fidelity than Taq DNA polymerase and Taq-based blends
Catalog No. 50-125-950
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.