Learn More
Alkali Scientific Alkali Scientific™ Hermle OLEUM Oil Testing Centrifuge, Z600-OL
SDP
Supplier:  Alkali Scientific Z600OL
Encompass
The Hermle Z600-OL OLEUM Oil-Testing Centrifuge is our stand alone oil testing centrifuge for pear shaped or conical ASTM oil testing tubes. A 4-place swing out rotor reaches up to 3,000rpm, or 2,213 xg to determine the water content of your oil sample. The OLEUM is equipped with a maintenance-free induction motor, stainless steel chamber, motor driven lid lock, and active imbalance detection and shutdown. Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf. In addition to retrieving saved programs, the easy to use front panel can adjust accel/decel rates, and change the end of run alarm tone.
Catalog No. 50-302-9574
Only null left
Add to Cart
Request a Quote