Learn More
Medchemexpress LLC HY-12054 10mg , Hesperadin CAS:422513-13-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1205410MG
Encompass_Preferred
HY-12054 10mg Hesperadin CAS: 422513-13-1 Hesperadin is an ATP competitive indolinone inhibitor of Aurora A and B. Hesperadin inhibits Aurora B with an IC50 of 250 nM. Hesperadin inhibits the growth of Trypanosoma brucei by blocking nuclear division and cytokinesis. Hesperadin also is a broad-spectrum influenza antiviral. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9977