Learn More
Apexbio Technology LLC Hesperadin 422513-13-1 50mg
SDP
Supplier:  Apexbio Technology LLC A411850
Encompass_Preferred
Hesperadin is a small-molecule ATP-competitive inhibitor targeting Aurora B kinase a serine/threonine-protein kinase involved in mitotic regulation By occupying the ATP-binding pocket of Aurora B and adjacent hydrophobic regions Hesperadin suppresses kinase activity with an IC50 approximately 250 nM In mitotic cells Aurora B-mediated phosphorylation of histone H3 Ser-10 is inhibited by Hesperadin this phosphorylation event is widely utilized as a mitotic biomarker Furthermore experimental applications of Hesperadin include investigating cell-cycle dynamics chromosome alignment and spindle checkpoint mechanisms as evidenced by its ability to disrupt proper chromosome alignment and segregation at approximately 40 nM IC50
Catalog No. 50-000-62562