Learn More
Selleck Chemical LLC Hesperadin 50mg 422513-13-1
SDP
Supplier:  Selleck Chemical LLC S152950MG
Encompass_Preferred
Hesperadin potently inhibits Aurora B with IC50 of 250 nM in a cell-free assay. It markedly reduces the activity of AMPK, Lck, MKK1, MAPKAP-K1, CHK1 and PHK while it does not inhibit MKK1 activity in vivo. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-820-5