Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hesperetin 7-O-glucoside | 31712-49-9 | 99.4% | 464.42 g/mol | C22H24O11 | 25MG
SDP

Supplier:  Medchemexpress LLC HY12513025MG

Encompass_Preferred

Hesperetin 7-O-glucoside is a flavanone monoglucoside used in biochemical and pharmacological research with reported anti-inflammatory and antihypertensive activity. It is suitable for studies of flavonoid metabolism and enzyme inhibition.

  • Chemical identity: C22H24O11, CAS 31712-49-9.
  • Molecular weight: 464.42 g/mol.
  • High purity: 99.4%.
  • Reported activities: anti-inflammatory and HMG-CoA reductase inhibition.
  • Supplied in small research quantities suitable for assay development and screening.

Catalog No. 50-002-37647


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.