Learn More
Medchemexpress LLC Hesperetin 7-O-glucoside | 31712-49-9 | MFCD20275042 | 99.4% | 464.42 g/mol | C22H24O11 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1251301MG
Encompass_Preferred
Hesperetin 7-O-glucoside is a flavanone glucoside derived from hesperetin, reported to have anti-inflammatory, antihypertensive, and antibacterial activity. It is used in biochemical and pharmacological research as an enzyme inhibitor and a metabolic probe.
  • Flavanone monoglucoside derived from hesperetin.
  • Used in studies of inflammation, lipid metabolism, and enzyme inhibition.
  • High purity suitable for analytical and research applications.
  • Solid, white to off-white powder in small research pack sizes.
  • Molecular weight 464.42 g/mol; formula C22H24O11.
  • Store protected from light; refrigerate short term, freeze in solvent for long term.
Catalog No. 50-002-35575