Learn More
Medchemexpress LLC Hesperidin | 520-26-3 | C28H34O15 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1533750MG
Encompass_Preferred
Hesperidin is a flavanone glycoside isolated from citrus fruits, known for its various biological properties. It decreases inflammatory mediators, exhibits significant antioxidant effects, and demonstrates antitumor and antiallergic activities.
  • Decreases inflammatory mediators
  • Exhibits significant antioxidant effects
  • Shows antitumor activity
  • Demonstrates antiallergic activities
  • Induces potent cytotoxic effects in human osteosarcoma MG-63 cells
  • Inhibits COX-2 and iNOS activity in LPS-activated RAW 264.7 cells
  • Suppresses MG-63 tumor growth in mice
Catalog No. 50-003-73378