Learn More
Medchemexpress LLC Hesperidin (Standard) | 520-26-3 | C₂₈H₃₄O₁₅ | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15337R50MG
Encompass_Preferred
Hesperidin (Standard) is the analytical standard for Hesperidin, intended for research and analytical applications. Also known as Hesperetin 7-rutinoside, this flavanone glycoside is isolated from citrus fruits and possesses numerous biological properties, including decreasing inflammatory mediators, significant antioxidant effects, and exhibiting antitumor and antiallergic activities. It serves as a reference standard for assays in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for hesperidin
  • Intended for research and analytical applications
  • Isolated from citrus fruits
  • Decreases inflammatory mediators
  • Significant antioxidant effects
  • Exhibits antitumor and antiallergic activities
  • Used as a reference standard in HPLC, GC, and MS
Catalog No. 50-003-72748