Learn More
Medchemexpress LLC Hesperidin (Standard) | 520-26-3 | C28H34O15 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1533710MM/1ML
Encompass_Preferred
Hesperidin, also known as Hesperetin 7-rutinoside, is a flavanone glycoside naturally isolated from citrus fruits. This compound is recognized for its various biological properties, including significant antioxidant effects, the ability to decrease inflammatory mediators, and demonstrated antitumor and antiallergic activities.
  • Decreases inflammatory mediators
  • Significant antioxidant effects
  • Exhibits antitumor activity
  • Exhibits antiallergic activity
  • Supports research in cancer
  • Targets apoptosis
  • Targets autophagy
  • Targets reactive oxygen species (ROS)
  • Functions as an endogenous metabolite
Catalog No. 50-003-72747