Learn More
Medchemexpress LLC 1-isothiocyanato-6-(methylsulfinyl)hexane | 4430-35-7 | MFCD03093720 | 99.2% | 205.34 g/mol | C8H15NOS2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1013715MG
Encompass_Preferred
Hesperin is an isothiocyanate compound found in Japanese horseradish (wasabi) that activates the Nrf2 antioxidant response pathway. It is supplied as a research-grade reagent with high purity for biochemical and cell-based assays investigating oxidative stress, cytoprotective signaling, and related pathways. Consult the safety data sheet for handling and storage instructions.
Catalog No. 50-193-2101