Learn More
Enzo Life Sciences HET-0016 (5mg). CAS: 339068-25-6
SDP
Supplier:  Enzo Life Sciences BMLEI3500005
Encompass_Preferred
HET-0016 is a potent and selective inhibitor of the biosynthesis of 20-HETE acting via inhibition of CYP4A. It inhibits cerebrovascular constriction induced by Ca2+ or arachidonate and inhibits angiogenesis and endothelial cell sprouting in various models. Alternative name: N-Hydroxy-N'-(4-butyl-2-methylphenyl)formamidine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (6 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0736