Learn More
Medchemexpress LLC HY-N2077 1mg Medchemexpress, Heteroclitin D CAS:140369-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20771MG
Encompass_Preferred
Medchemexpress, HY-N2077 1mg Heteroclitin D CAS:140369-76-2 Heteroclitin D is a lignin from Kadsura medicinal plants with anti-liqid peroxidation. Heteroclitin D inhibits L-type calcium channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2861