Learn More
Medchemexpress LLC HY-N1476 5mg Medchemexpress, Heterophyllin B CAS:145459-19-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14765MG
Encompass_Preferred
Medchemexpress, HY-N1476 5mg Heterophyllin B CAS:145459-19-4 Heterophyllin B is an active cyclic peptide isolated from Pseudostellaria heterophylla. Heterophyllin B provides a novel strategy for the treatment of esophageal cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1446