Learn More
Grainger Hex Bushing Forged Steel 2 x 1 in
Supplier:  Grainger 14J774
Encompass_Preferred
The Anvil Hex Bushing Forged Steel 14J774 is a hex bushing made from forged steel with a black oxide finish. It has an inside diameter of 1/2", an outside diameter of 7/8", and is 1-1/4" long. The bushing has a Rockwell hardness of C38-45 and is rated for temperatures up to 1000°F. It meets ASTM A108 standards and is manufactured in the USA. The bushing features a hex shape on the outside diameter for wrenching and has a smooth inside diameter hole. It can be used for a variety of mechanical applications requiring a hardened steel bushing.
Catalog No. 50-275-0955