Learn More
Grainger Hex Coupling 316 SS 3/4 x 1/2 in FNPT
Supplier:  Grainger 14M052
Encompass_Preferred
The Parker Hex Coupling 316 Stainless is a hex bore coupling made of 316 stainless steel. It has a 1-1/16 inch bore size and is rated for pressures up to 5000 psi. This coupling has a threaded female NPT connection on both ends and is designed for use in hydraulic applications where corrosion resistance is needed. The hex shape allows for easy wrench tightening during installation. Overall this is a high pressure stainless steel hydraulic coupling for corrosive environments.
Catalog No. 50-275-2737