Learn More
Med Vet International Hex Screw Driver, w/ Sleeve, J0412A, 2.0mm, Each
Supplier: Med Vet International JOR412A
Screwdrivers with screwholding sleeves hold the screw until it has engaged in the threaded hole. The sleeve slides back up the shaft to release the screw without disengaging it.