Learn More
Medchemexpress LLC HEX3 | 688805-40-5 | 99.74% | 1035.21 | 5 MG

Supplier: Medchemexpress LLC HYP03025MG
HEX3 is a fragment of the adenoviral hexon, which is the major capsid protein of adenovirion and is comprised of three identical polypeptide chains.
- Solid, white to off-white appearance.
- Soluble in water at ≥ 100 mg/mL (96.60 mM).
- Sequence: Lys-Tyr-Ser-Pro-Ser-Asn-Val-Lys-Ile.
- Suitable for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.