Learn More
Apexbio Technology LLC Hexa-D-arginine TFA 958204-56-3 1mg
SDP
Supplier:  Apexbio Technology LLC B56921
Encompass_Preferred
Hexa-D-arginine TFA (CAS 958204-56-3) is a peptide-derived inhibitor targeting furin-family proprotein convertases including furin PACE4 and PC1 It is designed to inhibit the proteolytic activity of these enzymes thereby interfering with the activation of precursor proteins Hexa-D-arginine TFA exerts its biological activity through competitive inhibition of furin-like enzymes In in vitro and in vivo studies Hexa-D-arginine TFA demonstrates inhibitory activity with IC50 values of 106 nM for furin 580 nM for PACE4 and 13 2 M for PC1 Based on these pharmacological properties Hexa-D-arginine TFA holds research potential in the study of pathogen-host interactions toxin processing pathways and protease regulation mechanisms
Catalog No. 50-000-92623