Learn More
Medchemexpress LLC HY-P0294 5mg Medchemexpress, Hexa-His CAS:64134-30-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02945MG
Encompass_Preferred
Medchemexpress, HY-P0294 5mg Hexa-His CAS:64134-30-1 Hexa-His is a peptide consisting of 6 His residues, used as a metal binding site for the recombinant protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4102