Learn More
Medchemexpress LLC Hexa-His | 64134-30-1 | 99.8% | 840.85 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYP029410MM/1ML
Encompass_Preferred
Hexa-His (6X His Tag) is a widely used affinity tag made up of six histidine residues. It facilitates protein purification and analysis by binding to affinity chromatography media that contain transition metal ions like nickel (Ni2+) or cobalt (Co2+).
  • Used for protein purification.
  • Binds to affinity chromatography media.
  • Supports in vitro protein purification and analysis.
Catalog No. 50-004-24966