Learn More
Medchemexpress LLC Hexa-His TFA | 1456728-20-3 | 99.8% | 954.87 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0294A10MG
Encompass_Preferred
Hexa-His TFA is a synthetic peptide composed of six histidine residues, commonly utilized as a metal-binding site for recombinant proteins. This peptide facilitates protein purification and detection by enabling interaction with metal ions like nickel or cobalt, which are often immobilized on chromatography resins or detection reagents.
  • Consists of 6 histidine residues
  • Used as a metal-binding site for recombinant proteins
  • Facilitates protein purification
  • Aids in protein detection
Catalog No. 50-004-22787