Learn More
Medchemexpress LLC Hexa-L-tyrosine | 6934-38-9 | MFCD00133897 | 99.9% | 997.05 | C54H56N6O13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW1420151MG
Encompass_Preferred
Hexa-L-Tyrosine is a hexamer peptide composed of six L-tyrosine residues supplied as a research-grade amino acid derivative. It is provided with high purity and characterized identity, and is commonly used as a building block in peptide synthesis, mass spectrometry studies, and biochemical assays involving tyrosine-rich sequences.
  • High purity (99.9%) suitable for analytical and synthetic applications.
  • Characterized molecular weight 997.05 and formula C54H56N6O13 for confident identification.
  • Useful as a peptide synthesis building block and for mass spectrometry experiments.
  • Supplied in small quantities appropriate for method development and analytical testing.
  • Recommended sealed storage to minimize moisture and preserve stability.
Catalog No. 50-002-83802