Learn More
Cayman Chemical Hexa-N-acetylchItohexaose 1mg
SDP
Supplier:  Cayman Chemical 178641 MG
Encompass_Preferred
A hexamer of N-acetylglucosamine that functions as an elicitor in plants, inducing the expression of chitinases; a substrate of lysozyme; binds LysM domains on certain proteins, including an endopeptidase of T. thermophilus
Catalog No. 50-296-5054