Learn More
Medchemexpress LLC Hexaammineruthenium(II) chloride | 15305-72-3 | MFCD00011477 | 274.16 | [Ru(NH3)6]Cl2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0335731G
Encompass_Preferred
Hexaammineruthenium(II) chloride is a yellow crystalline coordination compound used as a redox probe and catalyst in chemical and electroanalytical applications. It has limited solubility in water and is commonly used in fluorescence and electrochemistry studies.
  • Acts as a stable redox probe for electroanalytical methods.
  • Serves as a catalyst for hydrogenation, isomerization, and oxidation reactions.
  • Provides a yellow crystalline solid form suitable for handling and storage.
  • Has defined composition [Ru(NH3)6]Cl2 and molecular weight 274.16 for reproducible results.
  • Maintains stability under recommended storage conditions to preserve activity.
Catalog No. 50-002-99857