Learn More
Medchemexpress LLC Hexaammineruthenium(II) chloride | 15305-72-3 | MFCD00011477 | 98.0% | 274.16 g/mol | [Ru(NH3)6]Cl2 | 25 MG

Supplier: Medchemexpress LLC 25MGHYW033573
Hexaammineruthenium(II) chloride is the inorganic salt [Ru(NH3)6]Cl2 commonly used as a research reagent and as a redox and fluorescent probe in laboratory studies. It is provided as a solid powder with typical high purity and defined storage recommendations for long-term stability.
- Chemical formula: [Ru(NH3)6]Cl2.
- Molecular weight: 274.16 g/mol (reported values may vary by source).
- Purity: 98% typical for listed product specifications.
- Form: powder; store at -20°C for long term or 4°C for shorter storage.
- Common applications: redox chemistry, electrochemical studies, fluorescence labeling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.