Learn More
Medchemexpress LLC Hexaammineruthenium(II) chloride | 15305-72-3 | MFCD00011477 | 99.9% | 274.16 g·mol⁻¹ | [Ru(NH3)6]Cl2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW03357350MG
Encompass_Preferred
Hexaammineruthenium(II) chloride is a yellow crystalline ruthenium coordination compound supplied as a solid for laboratory research (formula [Ru(NH3)6]Cl2; CAS 15305-72-3). It is used as a redox reagent, catalyst, and electrochemical probe in inorganic and materials chemistry, with suppliers reporting high research-grade purities.
  • Solid, yellow crystalline appearance suitable for laboratory handling.
  • High purity for research applications, commonly 98-99.9%.
  • Well-defined molecular formula [Ru(NH3)6]Cl2 and molecular weight ~274.16 g·mol⁻¹.
  • Functions as a one-electron redox mediator and catalyst in coordination and electrochemical studies.
  • Available in small pack sizes (mg to g) for laboratory use.
Catalog No. 50-002-99322