Learn More
Medchemexpress LLC Hexaconazole | 79983-71-4 | ≥98% | C14H17Cl2N3O | 10 G
SDP
Supplier:  Medchemexpress LLC HYA027810G
Encompass_Preferred
Hexaconazole is a synthetic triazole fungicide and a demethylation enzyme inhibitor. It effectively targets various fungi, particularly ascomycetes and basidiomycetes, disrupting their cell membranes by inhibiting ergosterol biosynthesis. This chemical is known to enhance the activity of superoxide dismutase (SOD) and peroxidase.
  • Targets a broad range of fungi
  • Disrupts fungal cell membranes
  • Inhibits ergosterol biosynthesis
  • Enhances SOD and peroxidase activity
Catalog No. 50-004-32205