Learn More
Cayman Chemical Hexadecanal, 629-80-1, 500 mg
SDP
Supplier:  Cayman Chemical 9001996500
Encompass_Preferred
Molecular Formula C16H32O, Purity ≥98%, Formula Weight 240.4, Synonyms: 1-Hexadecanal, Palmitaldehyde; The 16-carbon free fatty aldehyde analog of palmitic acid that, in conjunction with NAD+, acts as a substrate for hexadecanal:NAD+ oxidoreductase (fatty aldehyde dehydrogenase).
Catalog No. NC1330889