Learn More
Medchemexpress LLC Hexadecanoic acid, 2-oxiranylmethyl ester | 7501-44-2 | MFCD14636325 | 98.0% | 312.49 g/mol | C19H36O3 | 1 G

Supplier: Medchemexpress LLC HYW1275781G
Glycidyl palmitate is an epoxide-containing long-chain fatty ester used as a biochemical reagent in life-science research. It is employed as an organic building block and analytical standard, with identity confirmed by NMR and mass spectrometry and specified storage conditions for stability.
- Epoxide-containing long-chain fatty ester.
- Cas number 7501-44-2.
- Molecular weight 312.49 g/mol.
- Purity 98.0%.
- Appearance white to off-white solid.
- Storage: powder -20°C up to 3 years or 4°C up to 2 years.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.