Learn More
Cayman Chemical HexadecnamIde 500mg
SDP
Supplier:  Cayman Chemical 21086500 MG
Encompass_Preferred
A primary fatty acid amide that is derived from palmitic acid (C16:0) and belongs to a class of important cell signaling lipids
Catalog No. 50-298-5173