Learn More
Medchemexpress LLC Hexadecyltrimethylammonium tetrafluoroborate | 73257-08-6 | MFCD00144896 | >98.0% | 371.35 g·mol⁻¹ | C19H42BF4N | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW127695250MG
Encompass_Preferred
Hexadecyltrimethylammonium tetrafluoroborate is a quaternary ammonium salt used as a biochemical reagent and phase-transfer surfactant in laboratory research. It is typically supplied as a white to off-white solid (formula C19H42BF4N; molecular weight 371.35 g·mol⁻¹) in small pack sizes for analytical and synthetic work. Handle according to the SDS for quaternary ammonium compounds.
  • Quaternary ammonium surfactant for phase-transfer and interfacial chemistry.
  • Suitable for biochemical assays and general laboratory research.
  • Available in small pack sizes suitable for analytical work, including 250 mg.
  • Reported high purity for reproducible results; verify exact purity on the SDS.
  • White to off-white solid that is easy to weigh and store under recommended conditions.
Catalog No. 50-002-82603