Learn More
Medchemexpress LLC Hexaethylene glycol monobenzyl ether | 24342-68-5 | MFCD06797107 | 99.9% | 372.45 g/mol | C19H32O7 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW04265410G
Encompass_Preferred
BnO-PEG6-OH (hexaethylene glycol monobenzyl ether) is a benzyl-protected six-unit polyethylene glycol linker supplied as a high-purity solid for use in conjugation chemistry. It is commonly used as a non-cleavable spacer in the synthesis of antibody-drug conjugates (ADCs) and as a PEG-based linker in proteolysis targeting chimeras (PROTACs), providing flexibility and improved solubility between functional moieties.
  • Non-cleavable six-unit PEG linker suitable for conjugation chemistry.
  • High purity, suitable for research and synthesis applications.
  • Improves solubility and provides flexible spacer length.
  • Available in multiple package sizes, including 10 G.
  • Stable when stored at recommended temperatures for long-term use.
Catalog No. 50-002-04299