Learn More
Medchemexpress LLC Hexaflumuron | 86479-06-3 | MFCD01743935 | 99.7% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1848
Encompass_Preferred
Hexaflumuron is an insect growth regulator (IGR) that inhibits chitin synthesis during insect molting. It is intended for research and analytical applications.
  • Targets insecticide.
  • Used in various research areas.
  • Applicable for research and analytical applications.
  • Inhibits chitin synthesis during insect molting.
Catalog No. 50-004-13848