Learn More
TARGETMOL CHEMICALS INC HEXAHYDROCURCUMIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ02885MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hexahydrocurcumin is a selective orally active COX-2 inhibitor and inactive against COX-1. purity: 99%
Catalog No. 50-257-8802