Learn More
Abcam He x amethonium bromide, nicotinic receptor blocker, 100MG
Supplier:  Abcam AB120538100MG
Encompass_Preferred
MW 362.19 Da. Ganglionic blocker; antagonist at α₃-containing nicotinic acetylcholine receptors (autonomic ganglia).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7509