Learn More
Electron Microscopy Sciences (HMDS), Hexamethyldisilazane, Reagent, 999-97-3, 100mL
SDP
Supplier:  Electron Microscopy Sciences 16700
Encompass_Preferred
Molecular Formula C6H19NSi2, Molecular Weight 161.395, HDMS is for fast preparation of soft insect tissues for SEM. It is a good substitution for critical point drying.
Catalog No. 50-243-18