Learn More
Medchemexpress LLC Hexamethylene bisacetamide | 3073-59-4 | MFCD00008684 | 99.6% | 200.28 g/mol | C10H20N2O2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12428425MG
Encompass_Preferred
Hexamethylene bisacetamide (HMBA) is a small-molecule research chemical used as a cell differentiation inducer and selective bromodomain inhibitor. It induces tumor cell differentiation, inhibits cell proliferation, and has been reported to modulate signaling pathways including Notch1, Bcl-2, and p53. The compound is supplied in solid and solution formats for laboratory studies in cell biology, pharmacology, and cancer research.
  • Induces cell differentiation in vitro.
  • Modulates Notch1, Bcl-2, and p53 signaling pathways.
  • Reported antitumor and antiproliferative activity.
  • Available as solid powder and DMSO solution formats.
  • Suitable for cell biology and pharmacology research.
Catalog No. 50-002-38229