Learn More
Medchemexpress LLC Hexamethylphosphoramide | 680-31-9 | MFCD00008303 | 99.9% | 179.20 g/mol | C6H18N3OP | 500g
SDP
Supplier:  Medchemexpress LLC HYY1155500G
Encompass_Preferred
Hexamethylphosphoramide is an orally active polar aprotic solvent flame retardant additive and carcinogen Hexamethylphosphoramide undergoes cytochrome P-450-mediated N-demethylation to Formaldehyde Hexamethylphosphoramide induces DNA-protein crosslinks Hexamethylphosphoramide has been linked to nasal tumors (squamous cell carcinoma adenoid squamous cell carcinoma) squamous metaplasia rhinitis tracheitis and reversible and irreversible infertility[1][2][3][4][5][6][7]
Catalog No. 50-002-66969