Learn More
Apexbio Technology LLC Hexaminolevulinate hydrochloride 140898-91-5 100mg
SDP
Supplier:  Apexbio Technology LLC A4729100
Encompass_Preferred
Hexaminolevulinate hydrochloride (CAS 140898-91-5) is a porphyrin precursor employed as a photosensitizing agent It is designed to enhance visualization of malignant tissue by promoting selective accumulation of photoactive porphyrin derivatives within neoplastic cells Hexaminolevulinate hydrochloride exerts its biological activity primarily through intracellular metabolic conversion to fluorescent porphyrins which produce red fluorescence under specific excitation wavelengths and improve optical distinction between tumor and normal tissues during examination Based on these pharmacological properties hexaminolevulinate hydrochloride holds research potential in photodynamic diagnosis particularly for urothelial carcinoma and oncology-related visualization studies
Catalog No. 50-000-93469