Learn More
Medchemexpress LLC Hexane, 1-[2-(2-azidoethoxy)ethoxy]-6-chloro- | 2568146-55-2 | 95.0% | 249.74 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138470
Encompass_Preferred
Azido-PEG2-C6-Cl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, which can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-52444